Skip to Content
Merck
Phosphotungstic acid hydrate 99.995% trace metals basis (Purity excludes up to 300 ppm Si)

Phosphotungstic acid hydrate

Phosphotungstic acid hydrate 99.995% trace metals basis (Purity excludes up to 300 ppm Si)
Synonym(s):
Tungstophosphoric acid
Linear Formula:
H3[P(W3O10)4] · xH2O
CAS Number:
Molecular Weight:
2880.05 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique