Skip to Content
Merck
Phosphocreatine di(tris) salt ≥97% (enzymatic)

Phosphocreatine di(tris) salt

Phosphocreatine di(tris) salt ≥97% (enzymatic)
Synonym(s):
Creatine phosphate di(tris) salt, N-(Imino[phosphonoamino]methyl)-N-methylglycine
Empirical Formula (Hill Notation):
C4H10N3O5P · 2C4H11NO3
CAS Number:
Molecular Weight:
453.38

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Tissue
Showing 1-1 of 1
Compare
Product Number
Description
Pricing