Skip to Content
Merck
PhIP ≥95% (HPLC)

PhIP

PhIP ≥95% (HPLC)
Synonym(s):
2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine
Empirical Formula (Hill Notation):
C13H12N4
CAS Number:
Molecular Weight:
224.26

Products

Brand
Biological Source
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing