Skip to Content
Merck
Phenol solution BioReagent, Molecular Biology

Phenol solution

Phenol solution BioReagent, Molecular Biology
Synonym(s):
Tris-equilabrated Phenol
Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
EC Number:

Products

Product Category
Brand
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Technique