![(−)-Perillyl alcohol analytical standard](/deepweb/assets/sigmaaldrich/product/structures/282/498/b9a8d961-67ad-49bd-be7e-110784caacd6/640/b9a8d961-67ad-49bd-be7e-110784caacd6.png)
(−)-Perillyl alcohol
![(−)-Perillyl alcohol analytical standard](/deepweb/assets/sigmaaldrich/product/structures/282/498/b9a8d961-67ad-49bd-be7e-110784caacd6/640/b9a8d961-67ad-49bd-be7e-110784caacd6.png)
Synonym(s):
(S)-4-Isopropenyl-1-cyclohexenylmethanol, (S)-p-Mentha-1,8-dien-7-ol
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing