Skip to Content
Merck
Pentane-1-sulfonic acid sodium salt for ion pair chromatography LiChropur™

Pentane-1-sulfonic acid sodium salt

Pentane-1-sulfonic acid sodium salt for ion pair chromatography LiChropur™
Synonym(s):
Pentane-1-sulfonic acid sodium salt
Empirical Formula (Hill Notation):
C5H11NaO3S
CAS Number:
Molecular Weight:
174.19

Products

Brand
Formula Weight
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing