Skip to Content
Merck
Penicillinase from Bacillus cereus lyophilized, powder, white, ~13 U/mg

Penicillinase from Bacillus cereus

Penicillinase from Bacillus cereus lyophilized, powder, white, ~13 U/mg
Synonym(s):
β-Lactamase I from Bacillus cereus, β-Lactamase II, Cephalosporinase, Penicillin amido-β-lactam hydrolase
CAS Number:

Products

Product Category
Brand
Biological Source
Color
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Showing 1-2 of 2