
Pefloxacin mesilate dihydrate

Synonym(s):
Pefloxacin mesylate dihydrate, 3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-, monomethanesulfonate, dihydrate, Pefloxacine monomethanesulfonate dihydrate, Pefloxacinium methanesulfonate dihydrate
Linear Formula:
C17H20FN3O3 • CH4O3S • 2H2O
CAS Number:
Molecular Weight:
465.49
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing