
PD 123,319 di(trifluoroacetate) salt hydrate

Synonym(s):
S-(+)-1-[(4-(Dimethylamino)-3-methylphenyl)methyl]-5-(diphenylacetyl)-4,5,6,7-tetrahydro-1H-imidazo[4,5-c]pyridine-6-carboxylic acid di(trifluoroacetate) salt hydrate
Empirical Formula (Hill Notation):
C31H32N4O3 · 2C2HF3O2 · xH2O
CAS Number:
Molecular Weight:
736.66 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing