![Paclitaxel Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/805/469/1b79f0d2-bbd1-4e6c-b2eb-c1d44d8e180d/640/1b79f0d2-bbd1-4e6c-b2eb-c1d44d8e180d.png)
Paclitaxel Related Compound A
![Paclitaxel Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/805/469/1b79f0d2-bbd1-4e6c-b2eb-c1d44d8e180d/640/1b79f0d2-bbd1-4e6c-b2eb-c1d44d8e180d.png)
Synonym(s):
Cephalomannine, NSC 318735
Empirical Formula (Hill Notation):
C45H53NO14
CAS Number:
Molecular Weight:
831.90
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2