![Orotic acid monohydrate 97%](/deepweb/assets/sigmaaldrich/product/structures/339/128/e5de969d-e68b-4cf5-96b6-508f69f48f0b/640/e5de969d-e68b-4cf5-96b6-508f69f48f0b.png)
Orotic acid monohydrate
![Orotic acid monohydrate 97%](/deepweb/assets/sigmaaldrich/product/structures/339/128/e5de969d-e68b-4cf5-96b6-508f69f48f0b/640/e5de969d-e68b-4cf5-96b6-508f69f48f0b.png)
Synonym(s):
2,6-Dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid
Empirical Formula (Hill Notation):
C5H4N2O4 · H2O
CAS Number:
Molecular Weight:
174.11
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing