![Orientin primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/811/038/9e3f0627-d199-4dc3-8fda-824055e35244/640/9e3f0627-d199-4dc3-8fda-824055e35244.png)
Orientin
![Orientin primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/811/038/9e3f0627-d199-4dc3-8fda-824055e35244/640/9e3f0627-d199-4dc3-8fda-824055e35244.png)
Synonym(s):
8-β-D-Glucopyranosyl-3′,4′,5,7-tetrahydroxyflavone, Luteolin 8-C-β-D-glucopyranoside, Luteolin-8-glucoside, Lutexin
Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing