Ondansetron
Synonym(s):
(RS)-1,2,3,9-Tetrahydro-9-methyl-3-(2-methylimidazol-1-ylmethyl)carbazol-4-one
Empirical Formula (Hill Notation):
C18H19N3O
CAS Number:
Molecular Weight:
293.36
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing