![Omeprazole Impurity B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/295/992/5213140a-b238-4296-ad01-6009aa00576f/640/5213140a-b238-4296-ad01-6009aa00576f.jpg)
Omeprazole Impurity B
![Omeprazole Impurity B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/295/992/5213140a-b238-4296-ad01-6009aa00576f/640/5213140a-b238-4296-ad01-6009aa00576f.jpg)
Synonym(s):
2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulfinyl]-5-methoxy-1H-benzimidazole, 2-[[(3,5-Dimethyl-2-pyridinyl)methyl]sulfinyl]--6-methoxy-1H-benzimidazol, 4-Desmethoxy Omeprazole
Empirical Formula (Hill Notation):
C16H17N3O2S
CAS Number:
Molecular Weight:
315.39
Products
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing