
Okadaic acid ammonium salt from Prorocentrum concavum

Synonym(s):
OA
Empirical Formula (Hill Notation):
C44H68O13 · NH3
CAS Number:
Molecular Weight:
822.03
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing