![Octreotide Acetate Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/403/175/020d2cad-26a7-4c3b-9d18-c0260883a4c8/640/020d2cad-26a7-4c3b-9d18-c0260883a4c8.jpg)
Octreotide acetate
![Octreotide Acetate Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/403/175/020d2cad-26a7-4c3b-9d18-c0260883a4c8/640/020d2cad-26a7-4c3b-9d18-c0260883a4c8.jpg)
Synonym(s):
Octreotide acetate salt
Empirical Formula (Hill Notation):
C49H66N10O10S2 · xC2H4O2
CAS Number:
Molecular Weight:
1019.24 (free base basis)
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2