Skip to Content
Merck
N-[Tris(hydroxymethyl)methyl]acrylamide contains ≤7% KCl, 93%

N-[Tris(hydroxymethyl)methyl]acrylamide

N-[Tris(hydroxymethyl)methyl]acrylamide contains ≤7% KCl, 93%
Synonym(s):
NAT, TRIS-acrylamide
Linear Formula:
H2C=CHCONHC(CH2OH)3
CAS Number:
Molecular Weight:
175.18
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing