NovaSeptum® GO Bottle Sampling System
Synonym(s):
NovaSeptum® GO Bottle
Products
Product Category
Brand
Product Line
Agency
Quality Segment
Application
Bottle material
Vial Bottle Material
Sterility
Sterilization Method
Showing 1-1 of 1
Compare
Product Number
Description
Pricing