![Noroxycodone-D3 hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/413/302/a8e5fc0e-0058-42c6-bf5a-106f1648356a/640/a8e5fc0e-0058-42c6-bf5a-106f1648356a.png)
Noroxycodone-D3 hydrochloride solution
![Noroxycodone-D3 hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/413/302/a8e5fc0e-0058-42c6-bf5a-106f1648356a/640/a8e5fc0e-0058-42c6-bf5a-106f1648356a.png)
Synonym(s):
Noroxycodone-D3 hydrochloride solution
Empirical Formula (Hill Notation):
C17H16D3NO4·HCl
Molecular Weight:
340.82
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing