![Nonylphenol diethoxylate solution 5 μg/mL in acetone, technical, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/175/248/ca8059fb-db7c-4c84-a114-51ca026fad8c/640/ca8059fb-db7c-4c84-a114-51ca026fad8c.png)
Nonylphenol diethoxylate solution
![Nonylphenol diethoxylate solution 5 μg/mL in acetone, technical, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/175/248/ca8059fb-db7c-4c84-a114-51ca026fad8c/640/ca8059fb-db7c-4c84-a114-51ca026fad8c.png)
Synonym(s):
NP-2EO technical
Empirical Formula (Hill Notation):
C19H32O3
Molecular Weight:
308.46
EC Number:
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2