![Nonoxynol 9 Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/235/516/c36544a0-c144-472c-ab46-7a1a775f8cd7/640/c36544a0-c144-472c-ab46-7a1a775f8cd7.jpg)
Nonoxynol 9
![Nonoxynol 9 Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/235/516/c36544a0-c144-472c-ab46-7a1a775f8cd7/640/c36544a0-c144-472c-ab46-7a1a775f8cd7.jpg)
Synonym(s):
α-(4-Nonylphenyl)-ω-hydroxynona(oxyethylene), 2-(4-Nonylphenoxy)ethanol
Linear Formula:
C17H28O2
Products
Brand
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing