Skip to Content
Merck
Nisoldipine ≥98% (HPLC)

Nisoldipine

Nisoldipine ≥98% (HPLC)
Synonym(s):
(±)-Isobutyl methyl 1,4-dihydro-2,6-dimethyl-4-(o-nitrophenyl)-3,5-pyridinedicarboxylate, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid methyl 2-methylpropyl ester, Sular
Empirical Formula (Hill Notation):
C20H24N2O6
CAS Number:
Molecular Weight:
388.41
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing