![Nifedipine impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/118/712/7cb1d8a6-904f-48d2-b94a-2ceb7ddaa2ee/640/7cb1d8a6-904f-48d2-b94a-2ceb7ddaa2ee.png)
Nifedipine impurity B
![Nifedipine impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/118/712/7cb1d8a6-904f-48d2-b94a-2ceb7ddaa2ee/640/7cb1d8a6-904f-48d2-b94a-2ceb7ddaa2ee.png)
Synonym(s):
Dimethyl 2,6-dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylate
Empirical Formula (Hill Notation):
C17H16N2O5
CAS Number:
Molecular Weight:
328.32
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing