Skip to Content
Merck
Neuropeptide Y human ≥95% (HPLC), powder, synthetic

Neuropeptide Y human

Neuropeptide Y human ≥95% (HPLC), powder, synthetic
Synonym(s):
NPY, hNPY, Neuropeptide Y, Human
Empirical Formula (Hill Notation):
C189H285N55O57S
CAS Number:
Molecular Weight:
4271.68

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-2 of 2