Neohesperidin dihydrochalcone
Synonym(s):
1-(4-((2-O-[6-Deoxy-α-L-mannopyranosyl]-β-D-glucopyranosyl)oxy)-2,6-dihydroxyphenyl)-3-[3-hydroxy-4-methoxyphenyl]-1-propanone, NHDC
Empirical Formula (Hill Notation):
C28H36O15
CAS Number:
Molecular Weight:
612.58
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing