Skip to Content
Merck
NBT crystals, pkg of 5 g

NBT

NBT crystals, pkg of 5 g
Synonym(s):
Nitrotetrazolium Blue chloride, 2,2′-bis(4-Nitrophenyl)-5,5′-diphenyl-3,3′-(3,3′-dimethoxy-4,4′-diphenylene)ditetrazolium chloride, 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride], p-Nitro-Blue tetrazolium chloride, p-Nitrotetrazolium blue, NBT, Nitro BT
Linear Formula:
C40H30N10O6 · 2Cl
CAS Number:
Molecular Weight:
817.64

Products

Formula Weight
Manufacturer
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing