![N-Acetylmuramic acid ≥98% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/163/561/a51ee625-880d-475d-a7b3-f4e683b7df85/640/a51ee625-880d-475d-a7b3-f4e683b7df85.png)
N-Acetylmuramic acid
![N-Acetylmuramic acid ≥98% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/163/561/a51ee625-880d-475d-a7b3-f4e683b7df85/640/a51ee625-880d-475d-a7b3-f4e683b7df85.png)
Synonym(s):
(R)-2-(Acetylamino)-3-O-(1-carboxyethyl)-2-deoxy-D-glucose, 2-Acetamido-2-deoxy-3-O-(D-1-carboxyethyl)-D-glucopyranose, MurNAc, NAMA
Empirical Formula (Hill Notation):
C11H19NO8
CAS Number:
Molecular Weight:
293.27
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing