
N-(2,6-Dimethylphenyl)chloroacetamide

Synonym(s):
1-Chloroacetylamino-2,6-dimethylbenzene, 2-Chloro-2′,6′-acetoxylidide, 2-Chloro-2′,6′-dimethylacetanilide, 2-Chloro-N-(2,6-dimethylphenyl)acetamide, N-(2,6-Dimethylphenyl)-2-chloroacetamide, N-(2,6-Xylyl)chloroacetamide, N-Chloroacetyl-2,6-dimethylaniline, Chloroacetamido-2,6-xylidine
Linear Formula:
ClCH2CONHC6H3(CH3)2
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1