
N-(2,4-Dimethylphenyl)formamide

Linear Formula:
(CH3)2C6H3NHCHO
CAS Number:
Molecular Weight:
149.19
Products
Product Category
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing