Skip to Content
Merck
Mycophenolic acid-d3 solution 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Mycophenolic acid-d3 solution

Mycophenolic acid-d3 solution 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C17H17O6D3
CAS Number:
Molecular Weight:
323.36
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique