![Monoclonal Anti-VSV Glycoprotein antibody produced in mouse clone P5D4, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/402/174/96631d0a-fc10-45cf-9feb-9e5ace279f33/640/96631d0a-fc10-45cf-9feb-9e5ace279f33.jpg)
Monoclonal Anti-VSV Glycoprotein antibody produced in mouse
![Monoclonal Anti-VSV Glycoprotein antibody produced in mouse clone P5D4, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/402/174/96631d0a-fc10-45cf-9feb-9e5ace279f33/640/96631d0a-fc10-45cf-9feb-9e5ace279f33.jpg)
Synonym(s):
Monoclonal Anti-VSV Glycoprotein
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Conjugate
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing