Monoclonal Anti-CD16 antibody produced in mouse
Synonym(s):
Anti-FCGR3A, Anti-FcγRIII
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2