![Monoclonal Anti-β-Galactosidase antibody produced in mouse clone GAL-40, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/881/458/814c156b-2e77-4c8d-babd-a8846526382e/640/814c156b-2e77-4c8d-babd-a8846526382e.jpg)
Monoclonal Anti-β-Galactosidase antibody produced in mouse
![Monoclonal Anti-β-Galactosidase antibody produced in mouse clone GAL-40, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/881/458/814c156b-2e77-4c8d-babd-a8846526382e/640/814c156b-2e77-4c8d-babd-a8846526382e.jpg)
Synonym(s):
Monoclonal Anti-β-Galactosidase
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2