
Mometasone impurity d

Synonym(s):
(9β,11β,16α)-21-Chloro-9,11-epoxy-17-[(2-furanylcarbonyl)oxy]-16-methylpregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C27H29ClO6
CAS Number:
Molecular Weight:
484.97
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing