![Moexipril hydrochloride powder, ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/409/940/05771ee9-d7a2-4b12-9f05-12d7c75f38e3/640/05771ee9-d7a2-4b12-9f05-12d7c75f38e3.png)
Moexipril hydrochloride
![Moexipril hydrochloride powder, ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/409/940/05771ee9-d7a2-4b12-9f05-12d7c75f38e3/640/05771ee9-d7a2-4b12-9f05-12d7c75f38e3.png)
Synonym(s):
(3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-3-isoquinolinecarboxylic acid hydrochloride
Empirical Formula (Hill Notation):
C27H35N2O7Cl
CAS Number:
Molecular Weight:
535.03
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade