Misoprostol
Synonym(s):
(±)-15-Deoxy-(16RS)-16-hydroxy-16-methylprostaglandin E1 methyl ester
Empirical Formula (Hill Notation):
C22H38O5
CAS Number:
Molecular Weight:
382.53
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3