Skip to Content
Merck
Millicell® 12 well hanging cell culture inserts pore size 5.0 μm, translucent PET membrane, diam. 12 mm, Tissue Culture (TC)-treated surface, sterile

Millicell® 12 well hanging cell culture inserts

Millicell® 12 well hanging cell culture inserts pore size 5.0 μm, translucent PET membrane, diam. 12 mm, Tissue Culture (TC)-treated surface, sterile

Products

Product Category
Brand
Color
Binding Type
Diameter (mm)
Height (mm)
Surface Area cm²
Wells
Working Volume (ml)
Quality Segment
Color
Housing Material
Membrane Material
Package Size
Pore size (µm)
Sterility
Sterilization Method
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing