Micro particles based on polymethacrylate
Synonym(s):
2-Propenoic acid,2-methyl homopolymer, 2-methyl-2-propenoic acid homopolymer, Microparticles based on PMAA, Microparticles based on poly(methacrylic acid), Polymethacrylate
Products
Brand
Physical Form
Format
Quality Segment
Application
Particle Size (µm)
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing