Skip to Content
Merck
Methyltrioctylammonium bromide 97%

Methyltrioctylammonium bromide

Methyltrioctylammonium bromide 97%
Synonym(s):
Trioctylmethylammonium bromide
Linear Formula:
[CH3(CH2)6CH2]3N(Br)CH3
CAS Number:
Molecular Weight:
448.61

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing