
Methyl trifluoromethanesulfonate

Synonym(s):
Methyl trifluoromethanesulfonate
Empirical Formula (Hill Notation):
C2H3F3O3S
CAS Number:
Molecular Weight:
164.10
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing