![Methylprednisolone British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/382/912/fa088ce6-1abc-4e93-a4fb-1153517599f7/640/fa088ce6-1abc-4e93-a4fb-1153517599f7.png)
Methylprednisolone
![Methylprednisolone British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/382/912/fa088ce6-1abc-4e93-a4fb-1153517599f7/640/fa088ce6-1abc-4e93-a4fb-1153517599f7.png)
Synonym(s):
6α-Methylprednisolone, 11β,17α,21-Trihydroxy-6α-methyl-1,4-pregnadiene-3,20-dione, 6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, Medrol, Medrone
Empirical Formula (Hill Notation):
C22H30O5
CAS Number:
Molecular Weight:
374.47
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing