Skip to Content
Merck
Methyl N,N,N′,N′-tetraisopropylphosphorodiamidite 97%

Methyl N,N,N′,N′-tetraisopropylphosphorodiamidite

Methyl N,N,N′,N′-tetraisopropylphosphorodiamidite 97%
Linear Formula:
[[(CH3)2CH]2N]2POCH3
CAS Number:
Molecular Weight:
262.37

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing