Skip to Content
Merck
Methyl arachidate analytical standard

Methyl arachidate

Methyl arachidate analytical standard
Synonym(s):
Arachidic acid methyl ester, Methyl eicosanoate
Linear Formula:
CH3(CH2)18COOCH3
CAS Number:
Molecular Weight:
326.56
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing