Skip to Content
Merck
Methyl α-bromoisobutyrate ≥99.0% (GC)

Methyl α-bromoisobutyrate

Methyl α-bromoisobutyrate ≥99.0% (GC)
Synonym(s):
Methyl 2-bromo-2-methylpropionate
Linear Formula:
(CH3)2CBrCOOCH3
CAS Number:
Molecular Weight:
181.03
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing