Skip to Content
Merck
Methyl 2-octynoate analytical standard

Methyl 2-octynoate

Methyl 2-octynoate analytical standard
Synonym(s):
Methyl heptine carbonate, NSC 72098
Linear Formula:
CH3(CH2)4C≡CCO2CH3
CAS Number:
Molecular Weight:
154.21
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique