Skip to Content
Merck
Methyl 2-methoxybenzoate 99%

Methyl 2-methoxybenzoate

Methyl 2-methoxybenzoate 99%
Synonym(s):
Methyl o-anisate
Linear Formula:
CH3OC6H4CO2CH3
CAS Number:
Molecular Weight:
166.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing