Skip to Content
Merck
Methimazole Related Compound C pharmaceutical secondary standard, certified reference material

Methimazole Related Compound C

Methimazole Related Compound C pharmaceutical secondary standard, certified reference material
Synonym(s):
1-Methyl-2-(methylthio)-1H-imidazole, 1-Methyl-2-(methylsulphanyl)-1H-imidazole
Empirical Formula (Hill Notation):
C5H8N2S
CAS Number:
Molecular Weight:
128.20

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2