Skip to Content
Merck
Metalaxyl (S)-enantiomer certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Metalaxyl (S)-enantiomer

Metalaxyl (S)-enantiomer certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
N-(2,6-Dimethylphenyl)-N-(2-methoxyacetyl)-L-alanine
Empirical Formula (Hill Notation):
C15H21NO4
CAS Number:
Molecular Weight:
279.33

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade