Skip to Content
Merck
MES sodium salt

MES sodium salt

MES sodium salt
Synonym(s):
2-(N-Morpholino)ethanesulfonic acid sodium salt, 4-Morpholineethanesulfonic acid sodium salt, MES, Sodium Salt, ULTROL® Grade
Empirical Formula (Hill Notation):
C6H12NNaO4S
CAS Number:
Molecular Weight:
217.22
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Technique