Skip to Content
Merck
Meclizine Related Compound A pharmaceutical secondary standard, certified reference material

Meclizine Related Compound A

Meclizine Related Compound A pharmaceutical secondary standard, certified reference material
Synonym(s):
4-Chlorobenzhydrol
Linear Formula:
ClC6H4CH(C6H5)OH
CAS Number:
Molecular Weight:
218.68

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2